Learn More
Andwin Scientific SPOR-KLENZ STERILANT 1 GAL
Supplier: Andwin Scientific 21899864EA
Made with a mixture of peracetic acid, hydrogen peroxide, and acetic acid, the Spor-Klenz� Concentrated Sterilant/Disinfectant is a liquid sterilant/disinfectant that performs its task of disinfecting in quick 10 minutes. This product is ideal for procedures wherein a critically high-level microbial control is necessary. It is sold individually in a quantity of 1 gallon.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.