Learn More
Wilmad Labglass SPRAYER TLC COMP 250ML
Supplier:  Wilmad Labglass LG4590102
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Sprayer, TLC Reagent, Complete. Capacity: 250mL; Joint Size: [ST] 24/40. For spraying chromagenic reagents when using thin layer or paper chromatography. Complete unit consists of spray head, flask and LG-1004-108 plastic clamp.
Catalog No. 50-800-668
missing translation for 'QuantityLeft'