Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR 12813, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
SR 12813 is a pregnane X receptor (PXR) agonist (EC50 values are 200 and 700 nM for human and rabbit PXR respectively). Activates the farnesoid X receptor (FXR) at μM concentrations. Displays hypocholesterolemic activity via enhanced degradation of HMG-CoA reductase.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 504.53 |
Percent Purity | >99% |
Chemical Name or Material | SR 12813 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction