Learn More
TARGETMOL CHEMICALS INC SSR504734 HCL 5MG
Supplier:  TARGETMOL CHEMICALS INC T90597
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SSR504734 is an orally active selective and reversible inhibitor of human rat and mouse GlyT1 ( IC 50 =18 15 and 38 nM respectively). SSR504734 exhibits activity in schizophrenia anxiety and depression models [1]. purity: 98%
Catalog No. 50-257-9027
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.