Learn More
TARGETMOL CHEMICALS INC ST-1535 5MG
Supplier:  TARGETMOL CHEMICALS INC T28861
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ST 1535 is a potent and orally active antagonist of A2A adenosine receptor with antiparkinsonian activity and antitremorigenic effects. ST 1535 exhibits the potential for the research of Parkinson’s disease. purity: 99%
Catalog No. 50-258-0487
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.