Learn More
Molecular Toxicology ST QUAD PC, 5 plates/sleeve, Store in Refrigerator
Supplier: Molecular Toxicology 21200
For phenotype confirmation of S. typhimurium Ames strains. Tests for histidine auxotrophy, pKM101, & pAQ1 presence. Supplied w/ crystal violet discs. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs. If your laboratory uses a specifically modified formulation or packaging, contact us for a full assessment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.