Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stachyose Tetrahydrate, MP Biomedicals™

Catalog No. p-7236582 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
100 mg
500 mg
1 g

Stachyose Tetrahydrate is a prebiotic and functional oligosaccharide.

  • Promotes proliferation of lactic acid bacteria (LAB) by induction of α-galactosidase production to hydrolyze stachyose.

Chemical Identifiers

CAS 10094-58-3
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
InChI Key UQZIYBXSHAGNOE-XNSRJBNMSA-N
IUPAC Name (2S,3R,4S,5R,6R)-2-{[(2R,3R,4S,5R,6S)-6-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Quantity 1 mg

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.