Learn More
Thomas Scientific Gel Staining Box, Clear Polymethylpentene (PMP) Box, Polypropylene Cover
SDP
Supplier:  Thomas Scientific 1222X52
Encompass
Electrophoresis gels and membranes can be easily stained, destained, fixed and handled in these boxes. Select either clear PMP box for easier viewing or the economical polypropylene box. Drain (with LDPE plug) located flush with the bottom of the box minimizes disturbance of the gel during liquid drainage Boxes and covers are unaffected by temperatures from -70ºC to +70ºC and are resistant to acids, alkalis, and many solvents Inside dimensions: 12.7 x 12.7 x 5.1cm deep (5 x 5 x 2"); liquid capacity up to 750mL
Catalog No. 50-146-0086
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.