Learn More
MilliporeSigma™ Supelco™ Stainless Steel 1/16 in. Capillary Tubing, Length: 30 cm
Use to connect the column to the detector compartment in a capillary gas chromatographic setup
Supplier:  MilliporeSigma™ Supelco™ 56734U
 View more versions of this product
Catalog No. 11-100-5164
Add to Cart
Description
Description
Specifications
Specifications
Capillary Tubing
0.030 in.
30 cm
Gas Chromatography (GC)
1/16 in.
Stainless Steel
Product Suggestions
Product Suggestions