Learn More
Glass Expansion Standard Sample Tubing 1/16OD x 0.75mm ID x 10m long
Supplier:  Glass Expansion STT1675
Encompass_Preferred
Standard Sample Tubing 1/16OD x 0.75mm ID x 10m long. Ten meters (32.8 ft) of chemically resistant PFA sample tubing. Outside diameter of 1/16” and internal diameter of 0.75 mm.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2942
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.