Learn More
Electron Microscopy Sciences MAC X-Ray Fluorescence Single Standard Pb
Supplier: Electron Microscopy Sciences 80093PB
These standards are supplied as 1/4" (33 mm) diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training. Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials (some of which are harmful).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.