Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Starter™ pH Meter ST2200
SCI_ED

Catalog No. S05306 Shop All OHAUS Products
Click to view available options
Includes:
ST2200 Meter, Electrode Holder, ST320 Electrode, 4 pH Buffer Mini Kits (50 mL)

The Starter™ pH Meter ST2200 is an affordable benchtop pH Meter that clearly displays information with five distinctly marked keys, allowing users to operate with minimal training.

  • Equipped with a large 3.5 in. LCD display
  • Features a 99 item memory for measurements and last calibration
  • Features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple
  • Choose between just the ST2200 meter with electrode holder and a full kit including ST2200 meter, electrode holder, ST320 electrode, 4 pH buffer mini kits (50 mL)

Specifications

Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1.5
Data Management Details Datalog for Measurements and Last Calibration
Includes ST2200 Meter, Electrode Holder, ST320 Electrode, 4 pH Buffer Mini Kits (50 mL)
Measurement Modes Auto-stop, Continuous
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Calibration Points 1 to 3
Temperature Range (Metric) 0.0°C to 100.0°C
Display Type LCD, 3.5 in. Segment Matrix
Data Points 99 Sets
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.