Learn More
LifeSign Status Flu A and B Qualitative Test Kit
SDP
Catalog No. 23264783 Shop All Lifesign LLC Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy
LifeSign Status Flu A and B Qualitative Test Kit is an in vitro rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
  • Rapid diagnosis for early treatment
  • Detects H1N1 (Swine Flu)
Kit Contents:
  • 22 test kit (Includes two free tests for QC)
  • Innovative flip test design
  • Easy-to-read cassette
  • Positive results in 10 minutes
  • Pre-measured developer solution capsule for increased accuracy and ease of use
  • Flock nasal swabs included for better specimen collection
Specifications
Detectable Analytes
Influenza type A and type B antigens
Sample Type
Nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
Type
Status Flu A and B Qualitative Test Kit