Learn More
LifeSign Status Strep A Test Kit
Rapid immunochromatographic assay for the detection of group A streptococcal antigen directly from throat swab specimens
Supplier: LifeSign 34025
Specifications
0.987 | |
5 to 10 min. | |
External negative and positve contols | |
Throat Swab | |
96.2% |
25 Tests | |
Group A streptococcal antigen | |
Cassette | |
Immunochromatographic Assay | |
Non-waived |