Learn More
STEMCELL Technologies CellAdhere™ Collagen I-Coated, 24-Well Flat-Bottom Plate, Size: 50 Units
SDP
Supplier:  STEMCELL Technologies 1001055
Encompass_Preferred
CellAdhereᵀᴹ Collagen I-Coated, Flat-Bottom Plates are sterile, clear polystyrene plates. For optimal cell attachment, these plates have been coated with high-purity collagen I derived from rat tail tendon. These plates can be used with a variety of cell types and cell culture applications.
Catalog No. NC2188040
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.