Learn More
STEMCELL Technologies CellAdhere™ Collagen I-Coated, 96-Well Flat-Bottom Plate, Size: 50 Units
SDP
Supplier:  STEMCELL Technologies 1001056
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CellAdhereᵀᴹ Collagen I-Coated, Flat-Bottom Plates are sterile, clear polystyrene plates. For optimal cell attachment, these plates have been coated with high-purity collagen I derived from rat tail tendon. These plates can be used with a variety of cell types and cell culture applications.
Catalog No. NC2188041