Learn More
Cole-Parmer SHP-200-MP Stuart Three Position Stirring Hot Plate; 230 VAC, 50 Hz
Supplier:  Cole-Parmer 5160008
Encompass_Preferred
Three independent stirring or heating/stirring positions yet only measures 23-5/8"L x 10-5/8"W (60 x 27 cm)Powerful magnets for strong magnetic coupling and stirring capacityEasily accommodate a 2-L beaker in each positionAluminum/silicon alloy top plate offers even plate temperatureA flashing "Hot" warning light indicates temperature above 50°C3 year warrantyThree independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.Please Note: Effective June 2022, the Stuart Stirring Hot Plate Model number SB162-3 has now been rebranded Cole-Parmer under Model number SHP-200-MP. All the same great features and specifications, just with a new and improved look.
Catalog No. 50-196-0434
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.