Learn More
Azzota Corp Stirring Heating Mantle, Capacity: 500Ml , Maximum Temperature Is 716'F (380'C). Stir Speed: 0 To 1400Rpm
Supplier:  Azzota Corp SHM500220
Encompass_Preferred
Stirring Heating Mantle, Capacity: 500ml , Maximum temperature is 716'F (380'C). Stir speed: 0 to 1400rpm
Catalog No. 50-161-2999
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.