Learn More
Pelican Products Inc. STORM CASE W/FOAM BLACK
DIMENSIONSInterior (L×W×D)14.12 x 13.19 x 9.90 in (35.9 x 33.5 x 25.1 cm)Exterior (L×W×D)15.25 x 15.50 x 10.22 in (38.7 x 39.4 x 26 cm)MEASUREMENTSLid Depth3.20" (8.1 cm)Bottom Depth6.30" (16 cm)Total Depth9.5" (24.1 cm)Int Volume1.07 ft³ (0.03 m³)Padlock Hole Diameter0.37" (0.9 cm)WEIGHTSWeight With Foam10.25 lbs (4.6 kg)Weight Empty8.00 lbs (3.6 kg)Buoyancy68.80 lbs (31.2 kg)MATERIALSBody MaterialInjection Molded HPX™ high performance resinLatch MaterialValox - Polybutylene Terephthalate (PBT)Latch (Alt) MaterialXenoy - Polyester/Polycarbonate blendO-Ring MaterialEPDMPins MaterialStainless SteelPins (Alt) MaterialAluminumFoam Material1.3/1.35 polyesterPurge Body MaterialValox - Polybutylene Terephthalate (PBT)Purge Body (Alt) MaterialXenoy - Polyester/Polycarbonate blendPurge Vent MaterialGortex membraneTEMPERATURESMinimum Temperature-20° F (-29 ° C)Maximum Temperature140° F (60 ° C)APPROVALSCertificationsI