Learn More
Roylco™ Straws & Connectors™

- Durable plastic straws fit snuggly on the connectors, but are flexible so you can build arches and tunnels
- Appropriate for a wide age range and easy to clean
- Age: 4+
- Made in USA
Specifications
Grade
1+
Format
Kit
Includes
232 Straws, 168 Connectors, Guide
Quantity
400 Pieces
Languages
English, Spanish, French, German
Subject
STEM, Physics - Matter
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.