Learn More
Gold Biotechnology Inc Streptavidin Agarose Resin 10 mL
Supplier: Gold Biotechnology Inc S10510
Streptavidin Agarose Resin is a high affinity biotin-binding chromatography resin with streptavidin covalently bound to 6% crosslinked agarose beads and a biotin binding capacity of >120 nmol/ml of gel. Immobilized Streptavidin resin is suitable for gravity flow columns, spin columns, and FPLC methods. Immobilized Streptavidin can be used for affinity chromatography purifications by separating biotinylated proteins from nonbiotinylated proteins and for affinity purification of antigens when used with biotinylated antibodies. Streptavidin is a biotin-binding protein found in the culture broth of the bacterium Streptomyces avidinii. Streptavidin can bind to four moles of biotin per mole of protein and it shows extremely high affinity, approximately 10-15 kDa. Streptavidin lacks the carbohydrate side chains present on avidin and has an isoelectric point of 6.5 to avidin's 10, far closer to that at which most useful biological interactions occur.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.