Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Streptomycin Sulfate
SCI_ED

Catalog No. S25797 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS

3810-74-0

Molecular Formula

C42H84N14O36S3

Molecular Weight (g/mol)

1457.376

InChI Key

QTENRWWVYAAPBI-FFCQDDOVSA-N

Synonym

Streptomycin A Sulfate, Streptomycin Sesquisulfate

PubChem CID

124080941

IUPAC Name

2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(

SMILES

CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Type

Streptomycin Sulfate

Packaging

Solid

Quantity

5 g

Assay Percent Range

>99% w/v

Linear Formula

C42H84N14O36S3

Formula Weight

1457.58

Chemical Name or Material

Streptomycin Sulfate

ShelfLife

2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.