Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SU 16f, Tocris Bioscience™

Catalog No. p-4784605 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PDGFRβ inhibitor

Potent and selective platelet-derived growth factor receptor β (PDGFRβ) inhibitor (IC50 = 10 nM) that displays > 14-fold, > 229-fold and > 10000-fold selectivity over VEGFR2, FGFR1 and EGFR respectively. Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 = 0.11 μM).

Chemical Identifiers

CAS 251356-45-3
Molecular Formula C24H22N2O3
Molecular Weight (g/mol) 386.451
InChI Key APYYTEJNOZQZNA-UHFFFAOYSA-N
Synonym su 16f, su-16f, 3-2,4-dimethyl-5-2-oxo-6-phenyl-1,2-dihydroindol-3-ylidenemethyl-1h-pyrrol-3-yl-propionic acid, 3-2,4-dimethyl-5-2-oxo-6-phenyl-1,2-dihydro-3h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid
PubChem CID 53396310
IUPAC Name 3-[2,4-dimethyl-5-[(2-oxo-6-phenyl-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(NC(=C1CCC(=O)O)C)C=C2C3=C(C=C(C=C3)C4=CC=CC=C4)NC2=O

Specifications

Quantity 10 mg
Formula Weight 386.44
Percent Purity >98%
Chemical Name or Material SU 16f
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.