Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SU 6668, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
ATP-competitive PDGFR, VEGF and FGFR inhibitor (IC50 values are 0.06, 2.43, 3.04 and > 100 μM at PDGFRβ, VEGFR2, FGFR1 and EGFR respectively). Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 values are 0.41, 9.3 and 16.5 μM for VEGF, FGF and PDGF-stimulated growth respectively) and induces > 75% growth inhibition against a broad range of tumor types in vivo. Exhibits antiangiogenic, anti-inflammatory, antimetastatic and proapoptotic activity and is orally active.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 310.35 |
Percent Purity | >99% |
Chemical Name or Material | SU 6668 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction