Learn More
World Precision Instrument SU-MPC385 Motorized Micromanipulator Systems-SU-MPC385-4
Supplier: World Precision Instrument SUMPC3854
The flagship in our line of precision micromanipulators, the motorized SU-MP285 is affordable yet offers advanced features found in manipulators costing thousands more. Custom engineered stepping motors, precision cross-roller bearing slides and proprietary worm gear capstan drives form the basis of the watch-like mechanical system. The controller provides power to the stage motors with a quiet linear power supply to minimize electrical noise radiation in your setup. Pipette holders and headstages are securely mounted to the SU-MP285 with one of our several unique and rigid mounting systems.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.