Learn More
Pfanstiehl Inc Sucrose, NF, EP, JP, ChP High Purity, Low Endotoxin, Beet Derived (S-124-2-MC) 100G GMP, Compendial CAS: 57-50-1
Sucrose is a non-reducing crystalline disaccharide made up of glucose and fructose, found in many plants but extracted as ordinary sugar mainly from sugar cane and sugar beets. Pfanstiehl, High Purity Low Endotoxin Sucrose is used in the biopharmaceutical industry to stabilize proteins, lipids, and carbohydrates throughout the formulation and freeze/thaw life cycle of therapeutics. It is also widely applied as a cryopreservative and media supplement in a variety of cell-based bioprocesses. It is preferred to trehalose in applications where solubility and/or viscosity issues are paramount.The benefits of sucrose include but are not limited to: Monoclonal Antibody(mAbs) stabilization Vaccines stabilization Peptide stabilization Antibody Drug Conjugate (ADC) stabilization Stem cell preservation Media SupplementationPfanstiehl Sucrose is available as NF (National Formulary), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia), and ChP (Chinese Pharmacopoeia).