Learn More
SuFu Polyclonal Antibody, Bethyl Laboratories

Description
The recommended shelf life for this product is 1 year from date of receipt.
SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).Specifications
SuFu | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UMX1, Q9Z0P7 | |
Sufu | |
Between 434 and 484 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SUFU | |
OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; suppressor of fused homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24069, 51684 | |
4° C | |
Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.