Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS Grade, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88716 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

5329-14-6

Molecular Formula

H3NO3S

Molecular Weight (g/mol)

97.09

InChI Key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

IUPAC Name

sulfamic acid

SMILES

NS(O)(=O)=O

Specifications

CAS Max %

1

Quantity

100 g

Grade

ACS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.