Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sulfur
SCI_ED

Catalog No. p-4416487 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Lab Grade - Flowers
Reagent Grade - Roll

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Color Yellow
Quantity 100 g
Linear Formula S
Formula Weight 32.06
Packaging Lab Grade - Flowers
Density 2.07g/mL
Chemical Name or Material Sulfur
ShelfLife 5 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.