Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.10 N, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88734 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

7664-93-9, 7732-18-5

Molecular Formula

H2O4S

Molecular Weight (g/mol)

98.07

MDL Number

MFCD00064589

InChI Key

QAOWNCQODCNURD-UHFFFAOYSA-N

IUPAC Name

sulfuric acid

SMILES

OS(O)(=O)=O

Specifications

Concentration

0.10 N

Quantity

60 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.