Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500mL
2.5 L
2.5L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
Glass Bottle, Single ship
PVC Coated Glass Bottle
Specifications
Specifications
Formula Weight | 98.08 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | PVC Coated Glass Bottle |
Specific Gravity | 1L = 1.84kg |
Quantity | 500mL |
Chemical Name or Material | Sulfuric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction