Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid/Phosphoric Acid, 1:1, Reagents

Catalog No. 01336202
Click to view available options
:

Sulfuric Acid/Phosphoric Acid, 1:1, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS

7664-93-9, 7664-38-2, 7664-38-2

Molecular Formula

H2O4S

Molecular Weight (g/mol)

98.07

MDL Number

MFCD00064589

InChI Key

QAOWNCQODCNURD-UHFFFAOYSA-N

IUPAC Name

sulfuric acid

SMILES

OS(O)(=O)=O

Specifications

pH

<1

Color

Colorless

Specific Gravity

1.7647

Quantity

1 L

Physical Form

Liquid

Grade

Reagent

Chemical Name or Material

Sulfuric Acid/Phosphoric Acid, 1:1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.