Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid-Silver Sulfate Reagent, for Chemical Oxygen Demand (COD), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
16 oz.
1 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 120 mL |
CAS Min % | 97.0, 0.53 |
CAS Max % | 100.0, 0.55 |
Color | Colorless |
Physical Form | Liquid |
Chemical Name or Material | Sulfuric Acid-Silver Sulfate Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction