Learn More
Enzo Life Sciences SUMO-1-AMC (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW00400050
Encompass_Preferred
Fluorogenic substrate for deSUMOylating enzymes. SUMO1-AMC is a sensitive fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760) and SENP2 (BML-UW9765). It is a particularly useful reagent for the study of deSUMOylating activity where detection sensitivity or continuous monitoring of activity is essential. Fluorogenic substrate for deSUMOylating enzymes. SUMO1-AMC is a sensitive fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760) and SENP2 (BML-UW9765). It is a particularly useful reagent for the study of deSUMOylating activity where detection sensitivity or continuous monitoring of activity is essential. Alternative name: Sentrin 1 AMC. Purity: ≥95% (HPLC). Formulation: Liquid. In 50mM HEPES, pH 7.5, containing 100 mM NaCl. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2225
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.