Learn More
CTL Scientific Supply Corp SUPHIDE TEST PAPER,PK/3 DISP
SDP
Supplier:  CTL Scientific Supply Corp 907613
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
SUPHIDE TEST PAPER - Pack of 3 dispensers each with a 5 meter roll of paper. The test paper allows the quick and easy detection of hydrogen sulphide. This paper does not contain lead acetate.
Catalog No. 50-117-3309
missing translation for 'QuantityLeft'