Learn More
Agilent Technologies SURE COMPETENT CELLS
SURE Competent Cells. For routine cloning of DNA with secondary structures, > /= 5 x 108 transformants/microgram. Restriction-Minus Competent Cells for Most Eukaryotic DNA. F- derivative for BAC cloning.