Learn More
TARGETMOL CHEMICALS INC SURFACTIN 10MG
Supplier:  TARGETMOL CHEMICALS INC T13041
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Surfactin is a potent cyclic lipopeptide biosurfactants that mediates flux of mono-and divalent cations such as calcium across lipid bilayer membranes with anti-bacterial anti-fungal antimycoplasma and hemolytic effects.
Catalog No. 50-257-7404
missing translation for 'QuantityLeft'