Learn More
Med Vet International 2.5G Ethicon Bone Wax, 12/Box
Supplier:  Med Vet International W31G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Ethicon's 2.5g bone wax contains a sterile mixture of beeswax, paraffin, and isopropyl palmitate. This product may be used for control of bleeding from bone surfaces. This does not act biochemically and is non-absorbable.
Catalog No. 50-252-2138