Learn More
Kingfisher Biotech Inc Swine Prolactin Receptor - 5ug
SDP
Supplier:  Kingfisher Biotech Inc RP2049S005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The Swine Prolactin Receptor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine Prolactin Receptor applications are for cell culture, ELISA standard, and Western Blot Control. Swine Prolactin Receptor yeast-derived recombinant protein can be purchased in multiple sizes. Swine Prolactin Receptor Specifications: (Molecular Weight: 24.5 kDa) (Amino Acid Sequence: SPPGKPEIFK CRSPEKETFT CWWKPGADGG LPTNYTLTYH KEGETFTHEC PDYKTGGPNS CYFNKKHTSI WTIYIITVNA TNQMGSSSSD PRYVDVTYIV EPDPPVNLTL ELKKPEDQKP YLWIKWLPPT LVDVRSGWLT LQYEIRLQPE KTAEWETHFA GQQTQFKILS LYPGQKYLVQ VRCKPDHGFW SEWSPESSIQ IPNDFSMKDT TM (212)) (Gene ID: 414916). For research use only. Made in the USA
Catalog No. 50-253-2506
missing translation for 'QuantityLeft'