Learn More
Trajan Scientific SYRINGE 100F 100UL FIXED
Supplier: Trajan Scientific 005000
SGE Series II Standard Fixed Needle Syringes, Volume: 100 uL, Needle Gauge: 25 G, Includes: Standard plunger, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Fixed, Diameter Outer: 0.31 in., 8 mm, Diameter Outer Needle: 0.01 in., 0.5 mm, Graduations: Major - 10.0 uL, Minor - 2.0 uL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.