Learn More
Creative Biomart Recombinant T.pallidum Partial p15 Antigen, GST-tagged
Supplier:  Creative Biomart TPP427
Encompass_Preferred
The E. coli derived recombinant protein is fused at N-terminus with GST tag and contains the Trp. Pallidum p15 immunodominant regions.
Catalog No. 50-208-4805
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.