Learn More
Creative Biomart Recombinant T.pallidum p17 Antigen, b-galactosidase-tagged
Supplier:  Creative Biomart TPP429
Encompass_Preferred
The E. coli derived recombinant protein contains the T. Pallidum p17 immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N- terminus.
Catalog No. 50-208-4807
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.