Learn More
Creative Biomart Recombinant T.pallidum TmpA Antigen, b-galactosidase-tagged
Supplier: Creative Biomart TMP435
The E. coli derived recombinant protein contains the T.Pallidum TmpA immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N-terminus.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.