Learn More
Clontech Labs 3P T-Vector pMD™19 (Simple), 1ug
SDP
Supplier:  Clontech Labs 3P 3271
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
T-Vector pMD19 (Simple) is a linearized vector with a single 3'-terminal thymidine at both ends. The T-overhang ends at the cloning site improve the efficiency of ligation of PCR products which contain A-overhangs at 3' -ends. The multiple cloning sites in the lac Z gene sequence have been deleted from this vector, but its β-galactosidase activity is not disrupted. Therefore, transformant colonies containing recombinant plasmids can be identified by blue/white screening. Due to lack of multiple cloning sites in the vector, the inserted PCR product cannot be recut using restriction enzymes. When the insert DNA fragment is recut from the vector, the PCR primers should be used to add a restriction site to the 5'-end for restriction enzyme digestion and subcloning. T-Vector pMD19 (Simple) is supplied with a Control Insert (500 bp) DNA for positive control reactions.
Catalog No. NC1591937
missing translation for 'QuantityLeft'