Learn More
G-Biosciences T4 DNA Polymerase
SCI_ED
T4 DNA Polymerase is used for 3'-overhang removal to form blunt ends, 5'-overhang fill-in to form blunt ends, probe labeling and DNA library preparation for Next-generation sequencing.
Supplier:  G-Biosciences 7862005
 View more versions of this product
Catalog No. S03110
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' - 3' direction with the presence of template and primer.
  • It has both DNA-dependent DNA polymerase activity and 3' - 5' exonuclease activity which make the enzyme useful for several molecular biology applications.
Specifications
Specifications
Gel Electrophoresis
Low
Standard
DNA Polymerase
750
Liquid
Standard PCR, End-point PCR, qPCR, Multiplex qPCR
Biotin and Other Hapten, No ROX, Biotin, Unlabeled, Unconjugated
Pathogen Detection, Non-Radioactive Labeling, DNA Sequencing, Next-Generation Sequencing (NGS), Cloning
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.