Learn More
G-Biosciences T4 GP32 Protein

- T4 GP32 is a single-stranded DNA binding protein with key role in DNA replication, recombination and repair.
- T4 GP32 binds cooperatively to single stranded DNA and stimulates cell-free DNA synthesis directed by the T4-encoded DNA polymerase.
Specifications
Quantity
100 μg
Product Type
Recombinant Protein
Conjugate
His-Tag, C-terminal His-Tag
Species
Human
Recombinant
Recombinant
Sterility
Sterile-filtered
Gender
Male
Form
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.