Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences T4 GP32 Protein

Click to view available options
Quantity:
100 μg
1000 μg
500 μg
Description
- T4 GP32 is a single-stranded DNA binding protein with key role in DNA replication, recombination and repair.
- T4 GP32 binds cooperatively to single stranded DNA and stimulates cell-free DNA synthesis directed by the T4-encoded DNA polymerase.
Specifications
Specifications
Quantity | 1000 μg |
Product Type | Recombinant Protein |
Conjugate | His-Tag, C-terminal His-Tag |
Species | Human |
Recombinant | Recombinant |
Sterility | Sterile-filtered |
Gender | Male |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction