Learn More
G-Biosciences T4 UvsY Protein

- During HR, the UvsX recombinase competes with the prebound single-stranded binding protein gp32 for DNA-binding sites and UvsY stimulates this filament nucleation event.
- UvsY stimulates the DNA-dependent ATPase activity of UvsX, lowers the critical concentration of UvsX that is required for activity, and promotes strand exchange.
- UvsY efficiently promotes the UvsX-catalyzed strand invasion reaction by recruiting to ssDNA–gp32 complexes, promotes the release of gp32, and favors the binding of UvsX.
Specifications
Quantity
100 μg
Product Type
Recombinant Protein
Conjugate
His-Tag, N-terminal His-Tag
Species
Human
Recombinant
Recombinant
Sterility
Sterile-filtered
Gender
Male
Form
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.