Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences T4 UvsY Protein

Click to view available options
Quantity:
100 μg
1000 μg
500 μg
Description
- During HR, the UvsX recombinase competes with the prebound single-stranded binding protein gp32 for DNA-binding sites and UvsY stimulates this filament nucleation event.
- UvsY stimulates the DNA-dependent ATPase activity of UvsX, lowers the critical concentration of UvsX that is required for activity, and promotes strand exchange.
- UvsY efficiently promotes the UvsX-catalyzed strand invasion reaction by recruiting to ssDNA–gp32 complexes, promotes the release of gp32, and favors the binding of UvsX.
Specifications
Specifications
Quantity | 100 μg |
Product Type | Recombinant Protein |
Conjugate | His-Tag, N-terminal His-Tag |
Species | Human |
Recombinant | Recombinant |
Sterility | Sterile-filtered |
Gender | Male |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction