Learn More
OHAUS™ Dry Block Heaters

Designed for applications that require repeatable results and superior temperature stability.
Supplier: OHAUS™ 30392080/EMD
Alert:
To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Description
Dry block heaters require either one or two modular blocks, sold separately.
- Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
Specifications
1.7 A | |
Ambient +5°C to 120°C | |
Ideal for incubation and activation of cultures, Enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedure | |
50/60 Hz | |
3.5 in. | |
92 in. Detachable, 3-wire cord and plug | |
39.1 cm | |
6.4 lb. | |
Accessory, Block |
Has 2 block capacity | |
120 V, 1.7 A, 210 w | |
Steel | |
50 min. | |
8.9 cm | |
15.4 in. | |
±0.1°C | |
2.9 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.